accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4650-60-6

IUPAC Name: 2-(isothiocyanatomethyl)furan
Synonyms: 2-(isothiocyanatomethyl)furan
Molecular Weight (g/mol): 139.17
Molecular Formula: C6H5NOS
InChi Key: ICXYINJACKJQQV-UHFFFAOYSA-N
SMILES: S=C=NCC1=CC=CO1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results