accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4651-81-4

IUPAC Name: methyl 2-aminothiophene-3-carboxylate
Synonyms: methyl 2-aminothiophene-3-carboxylate
Molecular Weight (g/mol): 157.19
Molecular Formula: C6H7NO2S
InChi Key: DGGJQLCAYQCPDD-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)SC=C1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results