accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4651-82-5

NH2SN

CAS RN 4651-82-5

IUPAC Name: 2-aminothiophene-3-carbonitrile
Synonyms: 2-aminothiophene-3-carbonitrile
Molecular Weight (g/mol): 124.16
Molecular Formula: C5H4N2S
InChi Key: XVGHZFWFGXDIOU-UHFFFAOYSA-N
SMILES: NC1=C(C=CS1)C#N
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)