accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4651-97-2

IUPAC Name: 2-amino-4-methylthiophene-3-carboxamide
Synonyms: 2-amino-4-methylthiophene-3-carboxamide
Molecular Weight (g/mol): 156.2
Molecular Formula: C6H8N2OS
InChi Key: AXLIJQRGPVJGSO-UHFFFAOYSA-N
SMILES: CC1=CSC(N)=C1C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results