accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 465514-13-0

IUPAC Name: 4-(morpholine-4-sulfonyl)benzene-1-sulfonyl chloride
Synonyms: 4-(morpholine-4-sulfonyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 325.78
Molecular Formula: C10H12ClNO5S2
InChi Key: ZZSBUDUHESWZJB-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)S(=O)(=O)N1CCOCC1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results