accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 466-37-5

IUPAC Name: tetraphenylethan-1-one
Synonyms: tetraphenylethanone
Molecular Weight (g/mol): 348.45
Molecular Formula: C26H20O
InChi Key: CFBBKHROQRFCNZ-UHFFFAOYSA-N
SMILES: O=C(C1=CC=CC=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)