accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOON

CAS RN 4664-08-8

IUPAC Name: 1H,3H-furo[3,4-c]pyridine-1,3-dione
Synonyms: furo[3,4-c]pyridine-1,3-dione
Molecular Weight (g/mol): 149.11
Molecular Formula: C7H3NO3
InChi Key: KFKMGUPDWTWQFM-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C2=CN=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results