accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHO

CAS RN 4677-18-3

IUPAC Name: 2-(oxan-4-yl)ethan-1-ol
Synonyms: 2-(oxan-4-yl)ethanol
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: XZXZZACRGBBWTQ-UHFFFAOYSA-N
SMILES: OCCC1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results