accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4677-20-7

IUPAC Name: 4-(2-bromoethyl)oxane
Synonyms: 4-(2-bromoethyl)oxane
Molecular Weight (g/mol): 193.08
Molecular Formula: C7H13BrO
InChi Key: WPDAWFCZGSQOPZ-UHFFFAOYSA-N
SMILES: BrCCC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results