accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4693-91-8

H3COClO

CAS RN 4693-91-8

IUPAC Name: 2-(4-methoxyphenyl)acetyl chloride
Synonyms: 2-(4-methoxyphenyl)acetyl chloride
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: CXJOONIFSVSFAD-UHFFFAOYSA-N
SMILES: COC1=CC=C(CC(Cl)=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)