accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4694-91-1

IUPAC Name: 6-nitro-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 6-nitro-3H-1,3-benzoxazol-2-one
Molecular Weight (g/mol): 180.12
Molecular Formula: C7H4N2O4
InChi Key: JGYJZHYTADCWIK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NC(=O)OC2=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results