CAS RN 470-37-1
CAS RN 470-37-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cinobufagine, TRC
CAS: 470-37-1 Molecular Formula: C26 H34 O6 Molecular Weight (g/mol): 442.54 Synonym: NSC 90325,Cinobufagin (6CI),14,15-Epoxy-14H-cyclopenta[a]phenanthrene, bufa-20,22-dienolide deriv.,14,15β-Epoxy-3β,16β-dihydroxy-5β-bufa-20,22-dienolide 16-Acetate,(3β,5β,15β,16β)-16-(Acetyloxy)-14,15-epoxy-3-hydroxy-bufa-20,22-dienolide IUPAC Name: [(1R,2S,4R,5R,6R,7R,10S,11S,14S,16R)-14-hydroxy-7,11-dimethyl-6-(6-oxopyran-3-yl)-3-oxapentacyclo[8.8.0.02,4.02,7.011,16]octadecan-5-yl] acetate SMILES: CC(=O)O[C@H]1[C@H]2O[C@]23[C@@H]4CC[C@@H]5C[C@@H](O)CC[C@]5(C)[C@H]4CC[C@]3(C)[C@H]1C6=COC(=O)C=C6