accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

IHNNO

CAS RN 470463-42-4

IUPAC Name: 5-iodo-2-(morpholin-4-yl)pyridin-1-ium
Synonyms: 5-iodo-2-(morpholin-4-yl)pyridin-1-ium
Molecular Weight (g/mol): 291.11
Molecular Formula: C9H12IN2O
InChi Key: RBMHFKCAFPUVRT-UHFFFAOYSA-O
SMILES: IC1=CC=C([NH+]=C1)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results