accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3CH3H3C

CAS RN 4706-90-5

IUPAC Name: 1,3-dimethyl-5-(propan-2-yl)benzene
Synonyms: 1-isopropyl-3,5-dimethylbenzene
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: RMKJTYPFCFNTGQ-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C)=CC(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results