CAS RN 4707-32-8
IUPAC Name:
2,2-dimethyl-2H,3H,4H,5H,6H-naphtho[1,2-b]pyran-5,6-dione
Synonyms:
β-lapachone lapachone lapachone, β-
Molecular Weight (g/mol):
242.27
Molecular Formula:
C15H14O3
InChi Key:
QZPQTZZNNJUOLS-UHFFFAOYSA-N
SMILES:
CC1(C)CCC2=C(O1)C1=CC=CC=C1C(=O)C2=O
1
–
3
of
3
results
2
beta-Lapachone (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
beta-Lapachone (>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.