CAS RN 471-34-1
Calcium carbonate, 99+%, ACS reagent
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, 98%
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 IUPAC Name: calcium carbonate SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, 98%, pure, light powder
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, 99%, extra pure
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, chelometric standard, ACS, 99.95-100.05%
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, ACS, 99.0% min
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium, AAS standard solution, Specpure™ Ca 1000μg/mL
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, 99%, for analysis, precipitated
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium, plasma standard solution, Specpure™ Ca 1000μg/mL
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, 98+%, pure, heavy powder
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, Puratronic™, 99.99% (metals basis)
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 IUPAC Name: calcium;carbonate SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, Puratronic™, 99.997% (metals basis)
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, ACS, low in alkalies, 99.0% min
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 IUPAC Name: calcium carbonate SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, 99+%, for biochemistry
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O
Calcium carbonate, 99.999%, (trace metal basis)
CAS: 471-34-1 Molecular Formula: CCaO3 Molecular Weight (g/mol): 100.09 MDL Number: MFCD00010906 InChI Key: VTYYLEPIZMXCLO-UHFFFAOYSA-L Synonym: calcium carbonate,limestone,chalk,calcite,carbonic acid calcium salt 1:1,marble,calofort u,aragonite,aeromatt,akadama PubChem CID: 10112 ChEBI: CHEBI:3311 SMILES: [Ca++].[O-]C([O-])=O