accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4717-96-8

IUPAC Name: 4-methyloxane
Synonyms: 4-methyloxane
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: OVRKATYHWPCGPZ-UHFFFAOYSA-N
SMILES: CC1CCOCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results