accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 472-61-7

IUPAC Name: (6S)-6-hydroxy-3-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-[(4S)-4-hydroxy-2,6,6-trimethyl-3-oxocyclohex-1-en-1-yl]-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl]-2,4,4-trimethylcyclohex-2-en-1-one
Synonyms: astaxanthin (3S,3'S)-astaxanthin all-trans-astaxanthin AstaREAL carophyll pink ovoester natupink astaxanthin, (3S,3'S)- E-astaxanthin astaxanthin (6CI)
Molecular Weight (g/mol): 596.85
Molecular Formula: C40H52O4
InChi Key: MQZIGYBFDRPAKN-UWFIBFSHSA-N
SMILES: C\C(\C=C\C=C(/C)\C=C\C1=C(C)C(=O)[C@@H](O)CC1(C)C)=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C(=O)[C@@H](O)CC1(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results