accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONCH3OH3C

CAS RN 4721-98-6

IUPAC Name: 6,7-dimethoxy-1-methyl-3,4-dihydroisoquinoline
Synonyms: 6,7-dimethoxy-1-methyl-3,4-dihydroisoquinoline
Molecular Weight (g/mol): 205.26
Molecular Formula: C12H15NO2
InChi Key: VASUQTGZAPZKFK-UHFFFAOYSA-N
SMILES: COC1=CC2=C(C=C1OC)C(C)=NCC2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results