accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4726-96-9

IUPAC Name: (2S)-2-azaniumyl-3-(benzyloxy)propanoate
Synonyms: (2S)-2-ammonio-3-(benzyloxy)propanoate
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: IDGQXGPQOGUGIX-VIFPVBQESA-N
SMILES: [NH3+][C@@H](COCC1=CC=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)