accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4727-72-4

OHN

CAS RN 4727-72-4

IUPAC Name: 1-benzylpiperidin-4-ol
Synonyms: 1-benzylpiperidin-4-ol
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: BPPZXJZYCOETDA-UHFFFAOYSA-N
SMILES: OC1CCN(CC2=CC=CC=C2)CC1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)