accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3PCH3CH3

CAS RN 4731-53-7

IUPAC Name: trioctylphosphane
Synonyms: trioctylphosphine
Molecular Weight (g/mol): 370.65
Molecular Formula: C24H51P
InChi Key: RMZAYIKUYWXQPB-UHFFFAOYSA-N
SMILES: CCCCCCCCP(CCCCCCCC)CCCCCCCC
Boiling Point 
  • (3)
  • (1)
Grade 
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)