accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

(R)(S)(S)(S)(R)(S)(R)(R)O(R)OH(R)OHHOHHHHMeMeMe

CAS RN 474-25-9

IUPAC Name: (4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Synonyms: chenodeoxycholic acid (+)-chenodeoxycholic acid anthropodesoxycholic acid anthropododesoxycholic acid chenocholic acid acid, gallodesoxycholic chenix chenodesoxycholic acid 3a,7a-dihydroxy-5b-cholanate acid, chenodeoxycholic
Molecular Weight (g/mol): 392.58
Molecular Formula: C24H40O4
InChi Key: RUDATBOHQWOJDD-BSWAIDMHSA-N
SMILES: C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)