accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4740-47-0

IUPAC Name: (2S)-2-azaniumyl-2-benzyl-3-hydroxypropanoate
Synonyms: (2S)-2-ammonio-2-benzyl-3-hydroxypropanoate
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: ZMNNAJIBOJDHAF-JTQLQIEISA-N
SMILES: [NH3+][C@](CO)(CC1=CC=CC=C1)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results