accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4740-78-7

IUPAC Name: 1,3-dioxan-5-ol
Synonyms: glycerol formal
Molecular Weight (g/mol): 104.11
Molecular Formula: C4H8O3
InChi Key: VCKSNYNNVSOWEE-UHFFFAOYSA-N
SMILES: OC1COCOC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)