accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHNOHCl

CAS RN 474010-60-1

IUPAC Name: 1-benzoyl-3-methylpiperazine hydrochloride
Synonyms: 1-benzoyl-3-methylpiperazine hydrochloride
Molecular Weight (g/mol): 240.73
Molecular Formula: C12H17ClN2O
InChi Key: OMFYILLDCYROOO-UHFFFAOYNA-N
SMILES: Cl.CC1CN(CCN1)C(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results