CAS RN 4741-99-5
IUPAC Name:
(2-aminoethyl)({3-[(2-aminoethyl)amino]propyl})amine
Synonyms:
1,4,8,11-tetraazaundecane 2,3,2-tetramine ethylenetrimethyleneethylenetetramine
Molecular Weight (g/mol):
160.27
Molecular Formula:
C7H20N4
InChi Key:
UWMHHZFHBCYGCV-UHFFFAOYSA-N
SMILES:
NCCNCCCNCCN