accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHCH3H3C

CAS RN 4747-21-1

IUPAC Name: methyl(propan-2-yl)amine
Synonyms: 2-propanamine, N-methyl-
Molecular Weight (g/mol): 73.14
Molecular Formula: C4H11N
InChi Key: XHFGWHUWQXTGAT-UHFFFAOYSA-N
SMILES: CNC(C)C
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)