accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4753-59-7

IUPAC Name: 4-bromobutyl acetate
Synonyms: 4-bromobutyl acetate
Molecular Weight (g/mol): 195.06
Molecular Formula: C6H11BrO2
InChi Key: UOABIRUEGSGTSA-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCBr
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)