accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4760-34-3

H3CNHH2N

CAS RN 4760-34-3

IUPAC Name: N1-methylbenzene-1,2-diamine
Synonyms: N1-methylbenzene-1,2-diamine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: RPKCLSMBVQLWIN-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results