accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHNOCH3OH3COCH3OCH3O

CAS RN 477-30-5

IUPAC Name: 3,4,5,14-tetramethoxy-10-(methylamino)tricyclo[9.5.0.0²,⁷]hexadeca-1(16),2,4,6,11,14-hexaen-13-one
Synonyms: (+-)-demecolcine colchamine kolchicin demecolcin substance F kolchamin colcemide deacetyl-N-methylcolchicine methylcolchicine deacetylmethylcolchicine
Molecular Weight (g/mol): 371.43
Molecular Formula: C21H25NO5
InChi Key: NNJPGOLRFBJNIW-UHFFFAOYNA-N
SMILES: CNC1CCC2=CC(OC)=C(OC)C(OC)=C2C2=CC=C(OC)C(=O)C=C12
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results