accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 477-73-6

IUPAC Name: 2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Synonyms: safranin
Molecular Weight (g/mol): 350.85
Molecular Formula: C20H19ClN4
InChi Key: QRYAEWIQIBAZOJ-UHFFFAOYSA-N
SMILES: [Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (12)
Quantity 
  • (1)
  • (4)
  • (6)
  • (2)
  • (2)
Color 
  • (2)
  • (2)
  • (3)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (6)
  • (3)
  • (1)
  • (3)
  • (12)
  • (1)
  • (4)
  • (6)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)