accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFBrO

CAS RN 477535-41-4

IUPAC Name: 3-bromo-5-(trifluoromethyl)benzaldehyde
Synonyms: 3-bromo-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 253.02
Molecular Formula: C8H4BrF3O
InChi Key: PCRLZGCXLNNMFL-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results