CAS RN 477535-43-6
IUPAC Name:
3-chloro-5-(trifluoromethyl)benzaldehyde
Synonyms:
3-chloro-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol):
208.56
Molecular Formula:
C8H4ClF3O
InChi Key:
NWSKKQLZBXTZTP-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC(Cl)=CC(C=O)=C1
1
–
1
of
1
results
1
3-Chloro-5-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
CAS: 477535-43-6 Molecular Formula: C8H4ClF3O Molecular Weight (g/mol): 208.56 MDL Number: MFCD04115845 InChI Key: NWSKKQLZBXTZTP-UHFFFAOYSA-N Synonym: 3-chloro-5-trifluoromethyl benzaldehyde,5-chloro-3-trifluoromethyl benzaldehyde,benzaldehyde, 3-chloro-5-trifluoromethyl,3-chloro-5-trifluoromethyl-benzaldehyde,pubchem18511,acmc-20a4r6 PubChem CID: 3857033 IUPAC Name: 3-chloro-5-(trifluoromethyl)benzaldehyde SMILES: FC(F)(F)C1=CC(Cl)=CC(C=O)=C1