accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOO

CAS RN 47758-37-2

IUPAC Name: 2-{[4-(9-{4-[(oxiran-2-yl)methoxy]phenyl}-9H-fluoren-9-yl)phenoxy]methyl}oxirane
Synonyms: 2-(4-{9-[4-(oxiran-2-ylmethoxy)phenyl]fluoren-9-yl}phenoxymethyl)oxirane
Molecular Weight (g/mol): 462.55
Molecular Formula: C31H26O4
InChi Key: LCSAOPVSVLGDLE-UHFFFAOYNA-N
SMILES: C(OC1=CC=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC=C(OCC2CO2)C=C1)C1CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results