accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 478-01-3

IUPAC Name: 2-(3,4-dimethoxyphenyl)-5,6,7,8-tetramethoxy-4H-chromen-4-one
Synonyms: nobiletin hexamethoxyflavone
Molecular Weight (g/mol): 402.4
Molecular Formula: C21H22O8
InChi Key: MRIAQLRQZPPODS-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C1=CC(=O)C2=C(OC)C(OC)=C(OC)C(OC)=C2O1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results