accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 47814-20-0

OOOOOOCCCHHHPrCF3CF3CF3CF3CF3CF3

CAS RN 47814-20-0

IUPAC Name: praseodymium(3+) 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
Synonyms: praseodymium(3+) 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 347.96
Molecular Formula: C5HF6O2Pr
InChi Key: GJZQTVLINUWRRY-UHFFFAOYSA-M
SMILES: [Pr+3].[O-]C(=CC(=O)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results