accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 479719-88-5

IUPAC Name: 2-{[2,2'-bithiophen]-5-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-{[2,2'-bithiophen]-5-yl}-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 292.22
Molecular Formula: C14H17BO2S2
InChi Key: HPOQARMSOPOZMW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(S1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results