accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOO

CAS RN 4799-68-2

IUPAC Name: 3-(benzyloxy)propan-1-ol
Synonyms: 3-(benzyloxy)propan-1-ol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: FUCYABRIJPUVAT-UHFFFAOYSA-N
SMILES: OCCCOCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)