accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaNaCH3H3CS(R)(R)NHOOOON(S)OO

CAS RN 4800-94-6

IUPAC Name: disodium (2S,5R,6R)-6-(2-carboxylato-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: geopen carbenicillin sodium pyopen carbecin sodium carbenicillin hyoper microcillin pyocianil carbenicillin disodium carbenicillin disodium salt
Molecular Weight (g/mol): 422.36
Molecular Formula: C17H16N2Na2O6S
InChi Key: RTYJTGSCYUUYAL-YCAHSCEMSA-L
SMILES: [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O
Quantity 
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (14)
  • (2)
Color 
  • (2)
  • (7)
  • (2)
Physical Form 
  • (2)
  • (6)
  • (3)
Type 
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (1)
  • (14)
  • (2)
  • (2)
  • (7)
  • (2)
  • (2)
  • (6)
  • (3)
  • (3)
  • (7)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)