accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 4805-22-5

IUPAC Name: 5,5'-dibromo-2,2'-bithiophene
Synonyms: 5,5'-dibromo-2,2'-bithiophene
Molecular Weight (g/mol): 324.05
Molecular Formula: C8H4Br2S2
InChi Key: SXNCMLQAQIGJDO-UHFFFAOYSA-N
SMILES: BrC1=CC=C(S1)C1=CC=C(Br)S1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (6)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)