accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOO

CAS RN 4808-48-4

IUPAC Name: diphenyl-2,5-dihydrofuran-2,5-dione
Synonyms: diphenylfuran-2,5-dione
Molecular Weight (g/mol): 250.25
Molecular Formula: C16H10O3
InChi Key: OUJCFCNZIUTYBH-UHFFFAOYSA-N
SMILES: O=C1OC(=O)C(=C1C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results