accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)(S)(S)CH3O(S)OOH3C

CAS RN 481-06-1

IUPAC Name: (3S,3aS,5aS,9bS)-3,5a,9-trimethyl-2H,3H,3aH,4H,5H,5aH,8H,9bH-naphtho[1,2-b]furan-2,8-dione
Synonyms: santonin (-)-α-santonin santoninic anhydride santoninum α-santonin semenen l-α-santonin santonin (substance) L-α-santonin (-)-santonine
Molecular Weight (g/mol): 246.31
Molecular Formula: C15H18O3
InChi Key: XJHDMGJURBVLLE-BOCCBSBMSA-N
SMILES: C[C@H]1[C@@H]2CC[C@@]3(C)C=CC(=O)C(C)=C3[C@H]2OC1=O
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results