accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 481-72-1

IUPAC Name: 1,8-dihydroxy-3-(hydroxymethyl)-9,10-dihydroanthracene-9,10-dione
Synonyms: aloe emodin emodine aloe-emodin 3-hydroxymethylchrysazin rhabarberone
Molecular Weight (g/mol): 270.24
Molecular Formula: C15H10O5
InChi Key: YDQWDHRMZQUTBA-UHFFFAOYSA-N
SMILES: OCC1=CC(O)=C2C(=O)C3=C(O)C=CC=C3C(=O)C2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results