accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OONH2S

CAS RN 4815-36-5

IUPAC Name: ethyl 2-amino-4-phenylthiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-phenylthiophene-3-carboxylate
Molecular Weight (g/mol): 247.31
Molecular Formula: C13H13NO2S
InChi Key: WYTHTMKMOSPACP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results