CAS RN 482-27-9
IUPAC Name:
4,9-dimethoxy-7H-furo[3,2-g]chromen-7-one
Synonyms:
isopimpinellin 5,8-dimethoxypsoralen 5,8-dimethoxypsoralene
Molecular Weight (g/mol):
246.22
Molecular Formula:
C13H10O5
InChi Key:
DFMAXQKDIGCMTL-UHFFFAOYSA-N
SMILES:
COC1=C2OC=CC2=C(OC)C2=C1OC(=O)C=C2
1
–
1
of
1
results
1
Isopimpinellin, TRC
CAS: 482-27-9 Molecular Formula: C13 H10 O5 Molecular Weight (g/mol): 246.22 Synonym: Isopimpinellin (6CI),4,9-Dimethoxy-7H-furo[3,2-g][1]benzopyran-7-one,5,8-Dimethoxy-6,7-furanocoumarin,5,8-Dimethoxypsoralen,5-Methoxyxanthotoxin,8-Methoxybergapten,Isoimpinellin,NSC 217988,NSC 401288 IUPAC Name: 4,9-dimethoxyfuro[3,2-g]chromen-7-one SMILES: COc1c2OC(=O)C=Cc2c(OC)c3ccoc13