accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 482-89-3

ONHHNO(Z)

CAS RN 482-89-3

IUPAC Name: (Z)-1H,1'H,3H,3'H-[2,2'-biindolylidene]-3,3'-dione
Synonyms: indigo blue
Molecular Weight (g/mol): 262.27
Molecular Formula: C16H10N2O2
InChi Key: COHYTHOBJLSHDF-YPKPFQOOSA-N
SMILES: O=C1\C(NC2=CC=CC=C12)=C1\NC2=CC=CC=C2C1=O

 No Refinements