accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PCl

CAS RN 48234-64-6

IUPAC Name: triphenyl(triphenylmethyl)phosphanium chloride
Synonyms: triphenyl(triphenylmethyl)phosphanium chloride
Molecular Weight (g/mol): 541.07
Molecular Formula: C37H30ClP
InChi Key: LZOHWIXYBMRNAP-UHFFFAOYSA-M
SMILES: [Cl-].C1=CC=C(C=C1)C(C1=CC=CC=C1)(C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results