accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3H3C

CAS RN 483-65-8

IUPAC Name: 1-methyl-7-(propan-2-yl)phenanthrene
Synonyms: reten retene
Molecular Weight (g/mol): 234.34
Molecular Formula: C18H18
InChi Key: NXLOLUFNDSBYTP-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C2C(C=CC3=C(C)C=CC=C23)=C1
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results