accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Cl

CAS RN 4830-93-7

IUPAC Name: (4-chlorobutyl)benzene
Synonyms: (4-chlorobutyl)benzene
Molecular Weight (g/mol): 168.66
Molecular Formula: C10H13Cl
InChi Key: FLLZCZIHURYEQP-UHFFFAOYSA-N
SMILES: ClCCCCC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results